missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-alpha-cyclohexylglycine, 98%

Product Code. 10336724 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10336724

Brand: Thermo Scientific Acros 399350010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109183-71-3 | C13H23NO4 | 257.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109183-71-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.32
MDL Number MFCD00133638
InChI Key QSUXZIPXYDQFCX-JTQLQIEISA-N
Synonym boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid
PubChem CID 7004938
IUPAC Name (2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O

Specifications

Melting Point 72°C to 76°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Specific Rotation +8° to +11° (20°C, 589nm) (c=1, CH3OH)
Quantity 1 g
Formula Weight 257.32
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material BOC-L-α-Cyclohexylglycine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.