missing translation for 'onlineSavingsMsg'
Learn More

Boc-L-aspartic Acid beta-Methyl Ester, TRC

Product Code. 30335693
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335693 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335693 Supplier Toronto Research Chemicals Supplier No. TRCB62133025G
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boc-L-aspartic Acid beta-Methyl Ester
CAS 59768-74-0
Molecular Formula C10 H17 N O6
Quantity 25 g
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-L-aspartic Acid 4-Methyl Ester, (S)-2-(tert-Butoxycarbonylamino)-4-methoxy-4-oxobutanoic Acid, N-tert-Butoxycarbonylaspartic Acid β-Methyl Ester
InChI Formula InChI=1S/C10H17NO6/c1-10(2,3)17-9(15)11-6(8(13)14)5-7(12)16-4/h6H,5H2,1-4H3,(H,11,15)(H,13,14)/t6-/m0/s1
SMILES COC(=O)C[C@H](NC(=O)OC(C)(C)C)C(=O)O
IUPAC Name (2S)-4-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Molecular Weight (g/mol) 247.245
Formula Weight 247.106
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.