missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals

Product Code. 11985501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11985501 25 g 25g
11945501 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11985501

Brand: Thermo Scientific Acros 440820250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30925-18-9
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.34
InChI Key LDRWTKQWSXGSTM-LBPRGKRZSA-N
Synonym boc-asp-obzl, boc-l-aspartic acid 1-benzyl ester, 1-benzyl n-boc-l-aspartate, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, commericial, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, pubchem12927, boc-l-asp h-obn
PubChem CID 7017887
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 96.0°C to 99.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Specific Rotation − 23.40
Formula Weight 323.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material BOC-L-Aspartic acid 1-benzyl ester
compliance-icons
Product Identifier
  • BOC-L-Aspartic acid 1-benzyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.