missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boc-Glu(OBzl)-OH, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Boc-Glu(OBzl)-OH |
| CAS | 13574-13-5 |
| Molecular Formula | C17H23NO6 |
| Quantity | 5 g |
| Synonym | N-(tert-Butoxycarbonyl)-L-glutamic Acid gamma-benzyl Ester, (S)-5-(Benzyloxy)-2-(tert-butoxycarbonylamino)-5-oxopentanoic Acid, (2S)-5-Benzyloxy-2-(tert-butoxycarbonylamino)-5-oxopentanoic Acid, (2S)-5-(Benzyloxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoic Acid, Nalpha-tert-Butoxycarbonyl-gamma-benzyl-L-glutamic Acidgamma-Benzyl N-(tert-butoxycarbonyl)glutamate |
| InChI Formula | InChI=1S/C17H23NO6/c1-17(2,3)24-16(22)18-13(15(20)21)9-10-14(19)23-11-12-7-5-4-6-8-12/h4-8,13H,9-11H2,1-3H3,(H,18,22)(H,20,21)/t13-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid |
| Molecular Weight (g/mol) | 337.37 |
| Formula Weight | 337.1525 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction