missing translation for 'onlineSavingsMsg'
Learn More

BOC-DL-Valine, 98%

Product Code. 10123194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123194 50 g Glass bottle 50g
10214674 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123194 Supplier Thermo Scientific Acros Supplier No. 425780500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54895-12-4 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54895-12-4
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00069973
InChI Key SZXBQTSZISFIAO-UHFFFAOYSA-N
Synonym boc-dl-valine, boc-dl-val-oh, n-boc-dl-valine, 2-tert-butoxycarbonyl amino-3-methylbutanoic acid, dl-n-tert-butoxycarbonyl valine, n-tert-butoxycarbonyl valine, 2-tert-butoxy carbonyl amino-3-methylbutanoic acid, n-tert-butoxycarbonyl-dl-valine, n-alpha-t-boc-l-valine
PubChem CID 270655
IUPAC Name 3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)C(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 112°C to 114°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(COOH)NHCOOC(CH3)3
Quantity 50 g
Formula Weight 217.26
Percent Purity 98%
Chemical Name or Material BOC-DL-Valine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.