missing translation for 'onlineSavingsMsg'
Learn More

Boc-D-serine, TRC

Product Code. 30340287
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30340287

Brand: Toronto Research Chemicals TRCB6673501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boc-D-serine
CAS 6368-20-3
Molecular Formula C8H15NO5
Quantity 1 g
Synonym N-[(1,1-dimethylethoxy)carbonyl]-D-serine, D-N-Carboxy-serine N-tert-Butyl Ester, (R)-2-(tert-Butoxycarbonylamino)-3-hydroxypropanoic Acid, (R)-2-(tert-Butoxycarbonylamino)-3-hydroxypropanoic Acid, N-Boc-D-serine, N-tert-Butoxycarbonyl-D-serine, tert-(Butoxycarbonyl)-D-serine
InChI Formula InChI=1S/C8H15NO5/c1-8(2,3)14-7(13)9-5(4-10)6(11)12/h5,10H,4H2,1-3H3,(H,9,13)(H,11,12)/t5-/m1/s1
SMILES CC(C)(C)OC(=O)N[C@H](CO)C(=O)O
IUPAC Name (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molecular Weight (g/mol) 205.21
Formula Weight 205.095
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.