missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Boc-D(OMe)-FMK Inhibitor

Product Code. 18136192 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18136192 1 mg 1mg
18126202 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18136192 Supplier R&D Systems™ Supplier No. NBP229395
 Restricted Item
Estimated Shipment: 07-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

A Novus product, part of R&D Systems. Applications: In vitro assay, In vivo assay, Block/Neutralize

TRUSTED_SUSTAINABILITY

Specifica

Color White
For Use With (Application) Blocking/Neutralizing, In vitro assay, In vivo assay
Content And Storage Store at -20°C. Avoid freeze-thaw cycles
Quantity 1 mg
Product Type Boc-D(OMe)-FMK Inhibitor
Molecular Weight (g/mol) 263
Product Line Novus
Inhibitors Boc-D(OMe)-FMK
Form Semi-solid

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.