missing translation for 'onlineSavingsMsg'
Learn More

BOC-D-FMK, MedChemExpress

Product Code. 16431329
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431329

Brand: Medchem Express HY13229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Boc-D-FMK is a cell-permeable, irreversible and broad spectrum caspase inhibitor. Boc-D-FMK inhibits apoptosis stimulated by TNF-α with an IC50 of 39 μM.

Boc-D-FMK is a cell-permeable, irreversible and broad spectrum caspase inhibitor. Boc-D-FMK inhibits apoptosis stimulated by TNF-α with an IC50 of 39 μM.

Specifications

Chemical Name or Material BOC-D-FMK
CAS 634911-80-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C11H18FNO5
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (379.85 mM; Need ultrasonic)
SMILES O=C(CC(C(CF)=O)NC(OC(C)(C)C)=O)OC
Molecular Weight (g/mol) 263.26
Formula Weight 263.26
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.