missing translation for 'onlineSavingsMsg'
Learn More

BOC-D-FMK, MedChemExpress

Product Code. 16475568
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475568 10 mg 10mg
16451737 50 mg 50mg
16448267 5 mg 5mg
16431329 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475568 Supplier Medchem Express Supplier No. HY13229
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Boc-D-FMK is a cell-permeable, irreversible and broad spectrum caspase inhibitor. Boc-D-FMK inhibits apoptosis stimulated by TNF-α with an IC50 of 39 μM.

Boc-D-FMK is a cell-permeable, irreversible and broad spectrum caspase inhibitor. Boc-D-FMK inhibits apoptosis stimulated by TNF-α with an IC50 of 39 μM.

Specifications

Chemical Name or Material BOC-D-FMK
CAS 634911-80-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C11H18FNO5
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (379.85 mM; Need ultrasonic)
SMILES O=C(CC(C(CF)=O)NC(OC(C)(C)C)=O)OC
Molecular Weight (g/mol) 263.26
Formula Weight 263.26
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.