missing translation for 'onlineSavingsMsg'
Learn More

Boc-Alpha-Methylalanine, TRC

Product Code. 30334284
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334284 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334284 Supplier Toronto Research Chemicals Supplier No. TRCB65694050G
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boc-alpha-Methylalanine
CAS 30992-29-1
Molecular Formula C9H17NO4
Quantity 50 g
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-2-methylalanine, N-Carboxy-2-methylalanine, N-tert-Butyl Ester, 2-(((tert-Butoxy)carbonyl)amino)-2-methylpropanoic Acid, 2-(Boc-amino)isobutyric Acid, 2-(tert-Butoxycarbonylamino)isobutanoic Acid, 2-(tert-Butoxycarbonylamino)isobutyric Acid, 2-(tert-Butoxycarboxylamino)-2-methylpropanoic Acid, 2-(tert-Butyloxycarbonylamino)isobutanoic Acid, 2-Methyl-2-((t-butyloxycarbonyl)amino)propionic Acid, 2-[(tert-Butoxycarbonyl)amino]-2-methylpropionic Acid, 208: PN: WO2004005478 PAGE: 71 Claimed Protein, Boc-2-methylalanine, N-(tert-Butoxycarbonyl)-2,2-dimethylglycine, N-(tert-Butoxycarbonyl)-alpha-methylalanine, N-(tert-Butyloxycarbonyl)-2-aminoisobutyric Acid, N-BOC-2-aminoisobutyric Acid, N-BOC-alpha-methylalanine, N-Boc-2,2-dimethylglycine, N-Boc-2-amino-2-methylpropanoic Acid, N-Boc-2-methylalanine, N-Boc-alpha,alpha-dimethylglycine, N-tert-Butoxycarbonyl-2-aminoisobutyric Acid, N-tert-Butoxycarbonyl-2-methylalanine, N-tert-Butoxycarbonyl-alpha-aminoisobutyric Acid, alpha-(Bocamino)isobutyric Acid
InChI Formula InChI=1S/C9H17NO4/c1-8(2,3)14-7(13)10-9(4,5)6(11)12/h1-5H3,(H,10,13)(H,11,12)
SMILES CC(C)(C)OC(=O)NC(C)(C)C(=O)O
IUPAC Name 2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molecular Weight (g/mol) 203.24
Formula Weight 203.1158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.