missing translation for 'onlineSavingsMsg'
Learn More

BOC-2-piperidylmethanol, 97%, Thermo Scientific™

Product Code. 11906391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11906391 10 g 10g
11976381 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11906391 Supplier Thermo Scientific Acros Supplier No. 437520100
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 157634-00-9
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
InChI Key PZTAGFCBNDBBFZ-UHFFFAOYSA-N
Synonym n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 2763853
IUPAC Name tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1CO

Specifications

Melting Point 74.4°C to 75.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 215.29
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material BOC-2-Piperidylmethanol
compliance-icons
Product Identifier
  • BOC-2-Piperidylmethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.