missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Bobcat339
Description
Bobcat339 is a selective cytosine-based inhibitor of ten-eleven translocation methylcytosine dioxygenase (TET) (IC50 values are 33 and 73 μM for TET1 and TET2, respectively). Bobcat339 reduces DNA 5-hydroxymethylcytosine (5hmC) levels in cells without inhibiting the DNA methyltransferase DNMT3a.
Specifications
Specifications
| Chemical Name or Material | 4-Amino-1-[1,1'-biphenyl]-3-yl-5-chloro-2(1H)-pyrimidinone |
| CAS | 2280037-51-4 |
| Quantity | 10 mg |
| Target | Other Oxidase/Oxygenase Compounds |
| Molecular Formula | C16H12ClN3O |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?