missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bobcat339

Product Code. 18004462
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18004462 50 mg 50mg
18014242 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 18004462

Brand: Tocris Bioscience™ 6977/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ten-eleven translocation methylcytosine dioxygenase (TET) inhibitor

Bobcat339 is a selective cytosine-based inhibitor of ten-eleven translocation methylcytosine dioxygenase (TET) (IC50 values are 33 and 73 μM for TET1 and TET2, respectively). Bobcat339 reduces DNA 5-hydroxymethylcytosine (5hmC) levels in cells without inhibiting the DNA methyltransferase DNMT3a.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-1-[1,1'-biphenyl]-3-yl-5-chloro-2(1H)-pyrimidinone
CAS 2280037-51-4
Quantity 50 mg
Target Other Oxidase/Oxygenase Compounds
Molecular Formula C16H12ClN3O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.