missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOB1, Mouse anti-Human, Clone: BOB1/2421, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79750-100UG
This item is not returnable.
View return policy
Description
BOB1 Monoclonal specifically detects BOB1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| BOB1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q16633 | |
| POU2AF1 | |
| Recombinant fragment (around aa 148-255) of human BOB1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| BOB1/2421 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5450 | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction