missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOB1 Antibody (BOB1/2425), Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £373.00
Specifications
| Antigen | BOB1 |
|---|---|
| Clone | BOB1/2425 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639546
|
Novus Biologicals
NBP2-75756-20ug |
20 μg |
£211.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628206
|
Novus Biologicals
NBP2-75756-100ug |
100 μg |
£373.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BOB1 Monoclonal antibody specifically detects BOB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| BOB1 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| 10 mM PBS with 0.05% BSA | |
| B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
| Recombinant fragment (around aa 148-255) of human BOB1 protein (exact sequence is proprietary) (Uniprot: Q16633) | |
| Primary | |
| Store at 4°C. |
| BOB1/2425 | |
| Western Blot 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q16633 | |
| 5450 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title