missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOB1 Antibody (BOB1/2421), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79885H
This item is not returnable.
View return policy
Description
BOB1 Monoclonal specifically detects BOB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BOB1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| POU2AF1 | |
| Recombinant fragment (around aa 148-255) of human BOB1 protein (exact sequence is proprietary) (Uniprot: Q16633) | |
| 0.1 mL | |
| Immunology | |
| Human | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| BOB1/2421 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction