missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOB-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587236
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.
Specifications
| BOB-1 | |
| Polyclonal | |
| Unconjugated | |
| POU2AF1 | |
| B-cell-specific coactivator OBF-1; BOB.1; Bob1; BOB-1; OBF.1; OBF1; Obf-1; OCAB; OCA-B; OCT-binding factor 1; POU class 2 associating factor 1; POU domain class 2-associating factor 1; POU domain, class 2, associating factor 1; Pou2af1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18985, 5450, 690528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16633, Q64693 | |
| POU2AF1 | |
| Recombinant full length Human POU2AF1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction