missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOB-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578115
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Positive Control: Jurkat, Raji, NCI-H929 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.
Spécification
| BOB-1 | |
| Polyclonal | |
| Unconjugated | |
| POU2AF1 | |
| B-cell-specific coactivator OBF-1; BOB.1; Bob1; BOB-1; OBF.1; OBF1; Obf-1; OCAB; OCA-B; OCT-binding factor 1; POU class 2 associating factor 1; POU domain class 2-associating factor 1; POU domain, class 2, associating factor 1; Pou2af1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q16633 | |
| POU2AF1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Human BOB1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu