missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BOB-1 Monoclonal Antibody (1170CT1.1.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537635
This item is not returnable.
View return policy
Description
BOB-1 Monoclonal Antibody for Western Blot
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.
Specifications
| BOB-1 | |
| Monoclonal | |
| Unconjugated | |
| POU2AF1 | |
| B-cell-specific coactivator OBF-1; BOB.1; Bob1; BOB-1; OBF.1; OBF1; Obf-1; OCAB; OCA-B; OCT-binding factor 1; POU class 2 associating factor 1; POU domain class 2-associating factor 1; POU domain, class 2, associating factor 1; Pou2af1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1170CT1.1.1 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q16633 | |
| POU2AF1 | |
| purified His-tagged POU2AF1 protein. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction