missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNTX maleate, Tocris Bioscience™
Standard δ1 selective antagonist
Chemical Identifiers
| CAS | 864461-31-4 |
|---|---|
| Molecular Formula | C31H31NO8 |
| Molecular Weight (g/mol) | 545.588 |
| InChI Key | GKHUUKGUNNYVGB-RAJBGOKPSA-N |
| Synonym | bntx maleate |
| PubChem CID | 124081000 |
| IUPAC Name | (4R,4aR,6E,7aR,12bR)-6-benzylidene-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;(E)-but-2-enedioic acid |
| SMILES | C1CC1CN2CCC34C5C(=O)C(=CC6=CC=CC=C6)CC3(C2CC7=C4C(=C(C=C7)O)O5)O.C(=CC(=O)O)C(=O)O |
Description
Standard selective δ1 opioid receptor antagonist. Also inhibits neurogenic ion transport mediated by a putative novel opioid receptor in porcine ileal mucosa.Chemical Identifiers
| 864461-31-4 | |
| 545.588 | |
| bntx maleate | |
| (4R,4aR,6E,7aR,12bR)-6-benzylidene-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;(E)-but-2-enedioic acid |
| C31H31NO8 | |
| GKHUUKGUNNYVGB-RAJBGOKPSA-N | |
| 124081000 | |
| C1CC1CN2CCC34C5C(=O)C(=CC6=CC=CC=C6)CC3(C2CC7=C4C(=C(C=C7)O)O5)O.C(=CC(=O)O)C(=O)O |
Specifications
| 864461-31-4 | |
| bntx maleate | |
| C1CC1CN2CCC34C5C(=O)C(=CC6=CC=CC=C6)CC3(C2CC7=C4C(=C(C=C7)O)O5)O.C(=CC(=O)O)C(=O)O | |
| 545.588 | |
| 545.59 | |
| BNTX maleate |
| C31H31NO8 | |
| GKHUUKGUNNYVGB-RAJBGOKPSA-N | |
| (4R,4aR,6E,7aR,12bR)-6-benzylidene-3-(cyclopropylmethyl)-4a,9-dihydroxy-1,2,4,5,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;(E)-but-2-enedioic acid | |
| 124081000 | |
| >99% |
Safety and Handling
Recommended Storage : Desiccate at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title