missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human N-Terminal Pro-BNP Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£166.00 - £400.00
Specifications
| Antigen | BNP |
|---|---|
| Clone | 2258B |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786848
![]() |
R&D Systems
MAB36042-SP |
25 μg |
£166.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18739568
![]() |
R&D Systems
MAB36042-100 |
100 μg |
£400.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for ELISA.Specifications
| BNP | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P16860 | |
| 4879 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2258B | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB | |
| E. coli-derived recombinant human Pro-BNP His27-His134 Accession # P16860 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Pro-BNP but does not detect mature human BNP peptide in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title