missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596084
This item is not returnable.
View return policy
Description
Positive Samples: Rat heart.
This gene is a member of the natriuretic peptide family and encodes a secreted protein which functions as a cardiac hormone. The protein undergoes two cleavage events, one within the cell and a second after secretion into the blood. The protein's biological actions include natriuresis, diuresis, vasorelaxation, inhibition of renin and aldosterone secretion, and a key role in cardiovascular homeostasis. A high concentration of this protein in the bloodstream is indicative of heart failure. Mutations in this gene have been associated with postmenopausal osteoporosis.
Specifications
| BNP | |
| Polyclonal | |
| Unconjugated | |
| Nppb | |
| 5 kDa cardiac natriuretic peptide; AA408272; Bnf; BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); BNP-32; BNP-34; Brain natriuretic factor; brain natriuretic peptide; Brain natriuretic peptide 32; Brain natriuretic peptide 45; brain type natriuretic peptide; gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide B; natriuretic peptide precursor B; natriuretic peptide precursor type B; natriuretic peptide type B; natriuretic peptides B; natriuretic protein; Nppb; type-B natriuretic factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18158, 25105, 4879 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide, pH 7.3 | |
| P13205, P16860, P40753 | |
| Nppb | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction