missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNP Antibody (NPPB/4493), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14020CL1
This item is not returnable.
View return policy
Description
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| BNP | |
| Monoclonal | |
| CoraFluor 1 | |
| BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
| Recombinant fragment of human BNP (exact sequence is proprietary) (Uniprot: P16860) | |
| 0.1 mL | |
| Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
| 4879 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| NPPB/4493 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction