Learn More
BNP Antibody (50E1cc), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB200-440C
386.75 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18261313
  • £486.00 / 0.20mL
Description
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BNP
Monoclonal
DyLight 650
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, ELISA
50E1cc
Western Blot, ELISA, Sandwich ELISA
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic human BNP, whole molecule, conjugated with carrier protein.
0.2 mL
Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
4879
Store at 4°C in the dark.
IgG1