Learn More
BNP Antibody (50E1cc), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB200-440AF647
Product Code. 18288324
  • £486.00 / 0.20mL
Description
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BNP
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, ELISA
50E1cc
Western Blot, ELISA, Sandwich ELISA
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic human BNP, whole molecule, conjugated with carrier protein.
0.2 mL
Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
4879
Store at 4°C in the dark.
IgG1