missing translation for 'onlineSavingsMsg'
Learn More

BNIPL Antibody (OTI9C1) [Biotin], Novus Biologicals™

Product Code. 30689150 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689150 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689150 Supplier Novus Biologicals Supplier No. NBP344726B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BNIPL Monoclonal antibody specifically detects BNIPL in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIPL
Applications Western Blot
Classification Monoclonal
Clone OTI9C1
Conjugate Biotin
Formulation PBS
Gene Alias bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein, BCL2/adenovirus E1B 19kD interacting protein like, BNIP-2 similar, BNIPl-1, BNIPL-2, BNIP-S, PP753
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-203 of human BNIPL(NP_612122) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 149428
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.