missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIPL Antibody (OTI8F6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | BNIPL |
|---|---|
| Clone | OTI8F6 |
| Dilution | Western Blot 1:500-2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306379
|
Novus Biologicals
NBP3-44929-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306143
|
Novus Biologicals
NBP3-44929-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BNIPL Monoclonal antibody specifically detects BNIPL in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| BNIPL | |
| Western Blot 1:500-2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein, BCL2/adenovirus E1B 19kD interacting protein like, BNIP-2 similar, BNIPl-1, BNIPL-2, BNIP-S, PP753 | |
| Human recombinant protein fragment corresponding to amino acids 1-203 of human BNIPL(NP_612122) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI8F6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cytokine Research | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 149428 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title