missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BNIP3L Recombinant Rabbit Monoclonal Antibody (K01_1N61)

Product Code. 30830420
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830420 50 μL
30826984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830420 Supplier Invitrogen™ Supplier No. MA564724
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

BNIP3L Recombinant Monoclonal Antibody for IHC (P), Western Blot

This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP3L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1N61
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene BNIP3L
Gene Accession No. O60238
Gene Alias Adenovirus E1B19K-binding protein B5; BCL2 interacting protein 3 like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa-interacting protein 3-like; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BCL2/adenovirus E1B 19kDa interacting protein 3-like; BCL2/adenovirus E1B 19kDa-interacting protein 3-like; BCL2/adenovirus E1B 19kD-interacting protein 3-like; BCL2/adenovirus E1B interacting protein 3-like; BNIP3A; BNIP3H; BNIP3L; C86132; D14Ertd719e; Nip3L; NIP3-like protein X; NIX
Gene Symbols BNIP3L
Host Species Rabbit
Immunogen A synthetic peptide of human BNIP3L.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 665
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.