missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNIP3L Recombinant Rabbit Monoclonal Antibody (7U2V9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558146
This item is not returnable.
View return policy
Description
Immunogen sequence: MSSHLVEPPP PLHNNNNNCE ENEQSLPPPA GLNSSWVELP MNSSNGNDNG NGKNGGLEHV PSSSSIHNGD MEKILLDAQH ESGQSSSRGS SHCDSPSPQE DGQIMFDVEM HTSRDHSSQS EEEVVEGEKE VEALKKSADW VSDWSSRPEN IPPKEFHFRH PKRSVSLSMR KSGAMKKGGI FSAEFLK.
This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
Specifications
| BNIP3L | |
| Recombinant Monoclonal | |
| 1.85 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9Z2F7 | |
| BNIP3L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-187 of human BNIP3L (NP_004322.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 7U2V9 | |
| Unconjugated | |
| BNIP3L | |
| Adenovirus E1B19K-binding protein B5; BCL2 interacting protein 3 like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa-interacting protein 3-like; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BCL2/adenovirus E1B 19kDa interacting protein 3-like; BCL2/adenovirus E1B 19kDa-interacting protein 3-like; BCL2/adenovirus E1B 19kD-interacting protein 3-like; BCL2/adenovirus E1B interacting protein 3-like; BNIP3A; BNIP3H; BNIP3L; C86132; D14Ertd719e; Nip3L; NIP3-like protein X; NIX | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12177 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction