missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNIP3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582532
This item is not returnable.
View return policy
Description
Immunogen sequence: SNGNDNGNGK NGGLEHVPSS SSIHNGDMEK ILLDAQHESG QSSSRGSSHC DSPSPQEDGQ IMFDVEMHTS RDHSSQSEEE VVEGEKEVEA LKKSADWVSD WSSRPENIPP KEFHFRHPKR SVSLSMRKSG AMK Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 96%.
This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
Specifications
| BNIP3L | |
| Polyclonal | |
| Unconjugated | |
| BNIP3L | |
| Adenovirus E1B19K-binding protein B5; BCL2 interacting protein 3 like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa-interacting protein 3-like; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BCL2/adenovirus E1B 19kDa interacting protein 3-like; BCL2/adenovirus E1B 19kDa-interacting protein 3-like; BCL2/adenovirus E1B 19kD-interacting protein 3-like; BCL2/adenovirus E1B interacting protein 3-like; BNIP3A; BNIP3H; BNIP3L; C86132; D14Ertd719e; Nip3L; NIP3-like protein X; NIX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60238 | |
| BNIP3L | |
| Recombinant protein corresponding to Human BNIP3L. Recombinant protein control fragment (Product #RP-90045). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction