missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNIP3L Polyclonal Antibody

Description
In Western blot, an upper band is detected at ~40kDa, which represents the monomer of Bnip3L. A suggested positive control is K562 cell lysate. PA5-19958 can be used with blocking peptide PEP-0083.
This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | BNIP3L |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BNIP3L |
| Gene Accession No. | O60238 |
| Gene Alias | Adenovirus E1B19K-binding protein B5; BCL2 interacting protein 3 like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa-interacting protein 3-like; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BCL2/adenovirus E1B 19kDa interacting protein 3-like; BCL2/adenovirus E1B 19kDa-interacting protein 3-like; BCL2/adenovirus E1B 19kD-interacting protein 3-like; BCL2/adenovirus E1B interacting protein 3-like; BNIP3A; BNIP3H; BNIP3L; C86132; D14Ertd719e; Nip3L; NIP3-like protein X; NIX |
| Gene Symbols | BNIP3L |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?