missing translation for 'onlineSavingsMsg'
Learn More

BNIP3L Mouse anti-Human, Clone: 1F6A3, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen BNIP3L
Clone 1F6A3
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17701179
View Documents
Proteintech
68118-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17791169
View Documents
Proteintech
68118-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BNIP3L Monoclonal antibody specifically detects BNIP3L in Human samples. It is validated for WB, ELISA
Specifications

Specifications

BNIP3L
1000 μg/mL
Monoclonal
Liquid
Human
665
Primary
Store at -20°C
1F6A3
Western Blot, ELISA
Unconjugated
Mouse
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
IgG1
Protein G purification
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.