missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNIP3L Monoclonal Antibody (ZN002)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 393300
This item is not returnable.
View return policy
Description
BNIP3L Monoclonal Antibody for Western Blot, ICC/IF, ELISA
This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
Specifications
| BNIP3L | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| O60238, Q9Z2F7 | |
| BNIP3L | |
| Synthetic peptide derived from the internal region of human, mouse and rat Nix proteins. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot, Immunocytochemistry | |
| ZN002 | |
| Unconjugated | |
| BNIP3L | |
| Adenovirus E1B19K-binding protein B5; BCL2 interacting protein 3 like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa-interacting protein 3-like; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BCL2/adenovirus E1B 19kDa interacting protein 3-like; BCL2/adenovirus E1B 19kDa-interacting protein 3-like; BCL2/adenovirus E1B 19kD-interacting protein 3-like; BCL2/adenovirus E1B interacting protein 3-like; BNIP3A; BNIP3H; BNIP3L; C86132; D14Ertd719e; Nip3L; NIP3-like protein X; NIX | |
| Mouse | |
| Protein A | |
| RUO | |
| 12177, 665 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction