missing translation for 'onlineSavingsMsg'
Learn More

BNIP3L Antibody, DyLight 680, Novus Biologicals™

Product Code. 18358056 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358056 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358056 Supplier Novus Biologicals Supplier No. NBP300002FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BNIP3L Polyclonal specifically detects BNIP3L in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP3L
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias Adenovirus E1B19K-binding protein B5, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, BCL2/adenovirus E1B 19-kd protein-interacting protein 3a, BCL2/adenovirus E1B 19kDa interacting protein 3-like, BNIP3a, BNIP3H, NIP3L, NIP3-like protein X, Nix, NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant BNIP3L (NP_004322.1; Ser2-Lys187).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Hypoxia, Mitophagy, p53 Pathway
Primary or Secondary Primary
Gene ID (Entrez) 665
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.