missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3L Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00002CL1
This item is not returnable.
View return policy
Description
BNIP3L Polyclonal antibody specifically detects BNIP3L in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| BNIP3L | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| Adenovirus E1B19K-binding protein B5, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, BCL2/adenovirus E1B 19-kd protein-interacting protein 3a, BCL2/adenovirus E1B 19kDa interacting protein 3-like, BNIP3a, BNIP3H, NIP3L, NIP3-like protein X, Nix, NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like | |
| Produced in rabbits immunized with purified, recombinant Human BNIP3L (Accession#: NP_004322.1; Ser2-Lys187). | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Hypoxia, Mitophagy, p53 Pathway | |
| 665 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction