missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596388
This item is not returnable.
View return policy
Description
Immunogen sequence: AFPPPAAEAH PAARRGLRSP QLPSGAMSQN GAPGMQEESL QGSWVELHFS NNGNGGSVPA SVSIYNGDME KILLDAQHES G Positive Samples: A375, Mouse heart; Cellular Location: Mitochondrion, Mitochondrion outer membrane, Single-pass membrane protein.
NIP3 is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. NIP3 contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein is known to induce apoptosis, even in the presence of BCL2.
Specifications
| BNIP3 | |
| Polyclonal | |
| Unconjugated | |
| BNIP3 | |
| BCL2 interacting protein 3; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3; BCL2/adenovirus E1B 19 kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B 19 kDa-interacting protein 3; BCL2/adenovirus E1B 19kDa interacting protein 3; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 3; Bnip3; NIP3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12176, 664, 84480 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.27 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O55003, Q12983 | |
| BNIP3 | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction