missing translation for 'onlineSavingsMsg'
Learn More

BNIP3 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18411047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411047 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411047 Supplier Novus Biologicals Supplier No. NBP177683MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BNIP3 Polyclonal antibody specifically detects BNIP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP3
Applications Flow Cytometry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human BNIP3 protein (within residues 50-150). [Swiss-Prot Q12983]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Hypoxia, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 664
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.