missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BNIP3 Polyclonal antibody specifically detects BNIP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | BNIP3 |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an internal region of the human BNIP3 protein (within residues 50-150). [Swiss-Prot Q12983] |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?