missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BNIP3 Monoclonal antibody specifically detects BNIP3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | BNIP3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 670621 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human BNIP3, Ser2-Glu160, Accession # Q12983 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?