missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92660-0.02ml
This item is not returnable.
View return policy
Description
BNIP2 Polyclonal antibody specifically detects BNIP2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| BNIP2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 2, BCL2/adenovirus E1B 19kDa interacting protein 2, BCL2/adenovirus E1B 19kD-interacting protein 2, BNIP-2, NIP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 125-280 of human BNIP2 (NP_004321.2). VELKEEWQDEDFPIPLPEDDSIEADILAITGPEDQPGSLEVNGNKVRKKLMAPDISLTLDPSDGSVLSDDLDESGEIDLDGLDTPSENSNEFEWEDDLPKPKTTEVIRKGSITEYTAAEEKEDGRRWRMFRIGEQDHRVDMKAIEPYKKVISHGGY | |
| 0.02 mL | |
| Apoptosis | |
| 663 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction