missing translation for 'onlineSavingsMsg'
Learn More

BNIP2 Antibody (8C6), Novus Biologicals™

Product Code. 18384628 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384628 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384628 Supplier Novus Biologicals Supplier No. H00000663M02100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BNIP2 Monoclonal antibody specifically detects BNIP2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 8C6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH02461
Gene Alias BCL2/adenovirus E1B 19 kDa protein-interacting protein 2, BCL2/adenovirus E1B 19kDa interacting protein 2, BCL2/adenovirus E1B 19kD-interacting protein 2, BNIP-2, NIP2
Host Species Mouse
Immunogen BNIP2 (AAH02461, 1 a.a. ~ 314 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEGVELKEEWQDEDFPIPLPEDDSIEADILAITGPEDQPGSLEVNGNKVRKKLMAPDISLTLDPSDGSVLSDDLDESGEIDLDGLDTPSENSNEFEWEDDLPKPKTTEVIRKGSITEYTAAEEKEDGRRWRMFRIGEQDHRVDMKAIEPYKKVISHGGYYGDGLNAIVVFAVCFMPESSQPNYRYLMDNLFKYVIGTLELLVAENYMIVYLNGATTRRKMPSLGWLRKCYQQIDRRLRKNLKSLIIVHPSWFIRTLLAVTRPFISSKFSQKIRYVFNLAELAELVPMEYVGIPECIKQVDQELNGKQDEPKNEQ
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 663
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.