missing translation for 'onlineSavingsMsg'
Learn More

BNC105, MedChemExpress

Product Code. 16499527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499527 5 mg 5mg
16446988 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499527 Supplier Medchem Express Supplier No. HY16114
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BNC105 is a tubulin polymerization inhibitor with potent antiproliferative and tumor vascular disrupting properties.

BNC105 is a tubulin polymerization inhibitor with potent antiproliferative and tumor vascular disrupting properties.

Specifications

Chemical Name or Material BNC105
CAS 945771-74-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H20O7
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (67.14 mM; Need ultrasonic)
SMILES COC1=CC(C(C2=C(C)OC3=C(O)C(OC)=CC=C23)=O)=CC(OC)=C1OC
Molecular Weight (g/mol) 372.37
Formula Weight 372.37
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.