missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BNC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566563
This item is not returnable.
View return policy
Description
Immunogen sequence: LSQEALESSED HFRAAYLLKD VAKEAYQDVA FTQQASQTSV IFKGTSRMGS LVYPITQVHS ASLESYNSGP LSEGTILDLS TTSSMKS Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Bnc 1 is a 994 amino acid transcription factor specific for squamous epithelium and for the constituent keratinocytes at a stage either prior to or at the very beginning of terminal differentiation. It is a zinc finger protein with three separated pairs of zinc fingers and a nuclear localization signal. Bnc 1 is a soluble protein that can shuttle between the nucleus and the cytoplasm, and its location depends on the proliferative potential of the cell. It is expressed relatively uniformly in the nucleoplasm, and phosphorylation on Ser-537 and Ser-541 leads to its cytoplasmic localization. It is present in the basal cell layer of the epidermis, in hair follicles and also in abundance in the germ cells of testis and ovary, and to a lower extent in thymus, spleen, mammary glands, placenta, brain and heart. Reports suggest that it plays a regulatory role in keratinocyte proliferation and also in rRNA transcription. It is also known to play a role in the differentiation of spermatozoa and oocytes.
Specifications
| BNC1 | |
| Polyclonal | |
| Unconjugated | |
| BNC1 | |
| AI047752; AW546376; basonuclin 1; BNC; bnc 1; Bnc1; BSN1; HsT19447; zinc finger protein basonuclin-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01954 | |
| BNC1 | |
| Recombinant Human BNC1. Recombinant protein control fragment (Product #RP-107218). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction