missing translation for 'onlineSavingsMsg'
Learn More

BNA Surrogate Standard Mixture 992 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30314461
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30314461 1 mL 1mL
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30314461

Brand: Dr. Ehrenstorfer DREGA09000992ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dichloroethane D4∣4-Bromofluorobenzene∣Toluene D8
CAS 17060-07-0, 460-00-4, 2037-26-5
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,2-Dichloroethane D4 1000 μg/mL∣4-Bromofluorobenzene 1000 μg/mL∣Toluene D8 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.