missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNA Internal Standard Mixture 997 2500 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1,4-Dichlorobenzene-D4∣Chlorobenzene D5∣Fluorobenzene |
| CAS | 3855-82-1, 3114-55-4, 462-06-6 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,4-Dichlorobenzene-D4 2500 μg/mL∣Chlorobenzene D5 2500 μg/mL∣Fluorobenzene 2500 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?