missing translation for 'onlineSavingsMsg'
Learn More

BN82002, MedChemExpress

Product Code. 16490987
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490987 50 mg 50mg
16484698 10 mg 10mg
16475526 100 mg 100mg
16427417 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490987 Supplier Medchem Express Supplier No. HY112776
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BN82002 is a potent, selective and irreversible inhibitor of CDC25 phosphatase family. BN82002 inhibits CDC25A, CDC25B2, CDC25B3, CDC25C CDC25A, and 25C-cat with IC50 values of 2.4, 3.9, 6.3, 5.4, and 4.6 μM, respectively. BN82002 displays ∼20-fold greater selectivity over CD45 tyrosine phosphatase.

BN82002 is a potent, selective and irreversible inhibitor of CDC25 phosphatase family. BN82002 inhibits CDC25A, CDC25B2, CDC25B3, CDC25C CDC25A, and 25C-cat with IC50 values of 2.4, 3.9, 6.3, 5.4, and 4.6 μM, respectively. BN82002 displays ∼20-fold greater selectivity over CD45 tyrosine phosphatase.

Specifications

Chemical Name or Material BN82002
CAS 396073-89-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H25N3O4
Quantity 50 mg
Solubility Information DMSO : ≥ 150 mg/mL (417.34 mM)
SMILES OC1=C(CN(C)CCC2=CC=C([N+]([O-])=O)C=C2)C=C(N(C)C)C=C1OC
Molecular Weight (g/mol) 359.42
Formula Weight 359.42
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.