missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMX Antibody (1C6), Novus Biologicals™
Mouse Monoclonal Antibody
£161.00
Specifications
| Antigen | BMX |
|---|---|
| Clone | 1C6 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
BMX Monoclonal specifically detects BMX in Human samples. It is validated for Western Blot, ELISA.Specifications
| BMX | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 660 | |
| Purified recombinant fragment of human BMX expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1C6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| BMX non-receptor tyrosine kinase, Bone marrow tyrosine kinase gene in chromosome X protein, EC 2.7.10, EC 2.7.10.2, Epithelial and endothelial tyrosine kinase, Etk/Bmx cytosolic tyrosine kinase, ETKcytoplasmic tyrosine-protein kinase BMX, NTK38, NTK38 tyrosine kinase, PSCTK2, PSCTK3 | |
| BMX | |
| IgG1 | |
| Unpurified | |
| 78 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title