Learn More
IKA™ BMT-50-S Tubes
Product Code. 11856810 Shop All IKA Products
Click to view available options
For Use With (Equipment):
BMT-50-S
Unit Size:
Pack of 10
Product Code. 11856810
Brand: IKA™ 0003699700
Only null left
Add to basket
Add to basket
Specifications
Product Type BMT-50-S Tube
Volume Range (Metric) 15 to 50 mL
For Use With (Application) Dry milling of dry and brittle samples, cell maceration, processing of materials mixed with fluids
For Use With (Equipment) BMT-50-S
Packaging Quantity 10/Pk.