missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ BMT-50-S-M-Gamma Tubes
Shop All IKA Products
Click to view available options
For Use With (Equipment):
BMT-50-S-M
Unit Size:
Pack of 10
Description
- Components: Tube, stainless steel balls, cover
- Additional media quantities may be added during operation
- Sampling during operation
- Sample preparation under sterile conditions
- Germ free conservation of samples (tissue, blood, etc.)
Specifications
Specifications
| Product Type | BMT-50-S-M Tube |
| Capacity (Metric) | 50 mL |
| For Use With (Application) | Introduction of media or sample extraction during the stirring, dispersing, or milling process under sterile conditions |
| For Use With (Equipment) | BMT-50-S-M |
| Packaging Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction