missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £359.00
Specifications
| Antigen | BMS1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633112
|
Novus Biologicals
NBP2-92629-0.02ml |
0.02 mL |
£153.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673422
|
Novus Biologicals
NBP2-92629-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMS1 Polyclonal antibody specifically detects BMS1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| BMS1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| BMS1 homolog, ribosome assembly protein (yeast), KIAA0187, ribosome assembly protein, ribosome assembly protein BMS1 homolog, ribosome biogenesis protein BMS1 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-900 of human BMS1 (NP_055568.3). LSAEEEDSENEEAIRKKLSKPSQVSSGQKLGPQNFIDETSDIENLLKEEEDYKEENNDSKETSGALKWKEDLSRKAAEAFLRQQQAAPNLRKLIYGTVTEDNEEEDDDTLEELGGLFRVNQPDRECKHKADSLDCSRFLVEAPHDWDLEEVMNSIRDCFVTGKWEDDKDAAKVLAEDEELYGDFEDLETGDVHKGKSGPNTQNEDIEKEVKEEIDPDEEESAKKKHLDKKRKLKEMFDAEYDEGESTYFDDLKGEMQKQAQLNRAEFEDQDDEARVQYEGFRPGMYVRIEIENVPCEFVQN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9790 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title